Learn More
Bioss KD-Validated Insulin receptor
SDP
Catalog No. 5000065142
Encompass_Preferred
Insulin Receptor Primary Antibody Unconjugated Public Immunogen Range 910-976/1382 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-65142 Supplier Bioss Supplier No. BSM90014R
Insulin Receptor Primary Antibody Unconjugated Public Immunogen Range 910-976/1382 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide