Learn More
Bioss KD-Validated Integrin subunit
SDP
Catalog No. 5000065126
Encompass_Preferred
Integrin Subunit Alpha V Primary Antibody Unconjugated Public Immunogen Range 31-250/1048 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-65126 Supplier Bioss Supplier No. BSM90029R
Integrin Subunit Alpha V Primary Antibody Unconjugated Public Immunogen Range 31-250/1048 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide