Learn More
Bioss KD-Validated Intraflagellar tr
SDP
Catalog No. 5000065185
Encompass_Preferred
Intraflagellar Transport 88 Primary Antibody Unconjugated Public Immunogen Range 1-28/824 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-65185 Supplier Bioss Supplier No. BSM90172R
Intraflagellar Transport 88 Primary Antibody Unconjugated Public Immunogen Range 1-28/824 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide