Learn More
Bioss KD-Validated ITPK1 Rabbit Reco
SDP
Catalog No. 5000066184
Encompass_Preferred
Inositol-Tetrakisphosphate 1-Kinase Primary Antibody Unconjugated Public Immunogen Range 362-402/414 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66184 Supplier Bioss Supplier No. BSM90378R
Inositol-Tetrakisphosphate 1-Kinase Primary Antibody Unconjugated Public Immunogen Range 362-402/414 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide