Learn More
Bioss KD-Validated KMT6 / EZH2 Rabbi
SDP
Catalog No. 5000066281
Encompass_Preferred
Enhancer Of Zeste 2 Polycomb Repressive Complex 2 Subunit Primary Antibody Unconjugated Public Immunogen Range 1-55/746 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66281 Supplier Bioss Supplier No. BSM90370R
Enhancer Of Zeste 2 Polycomb Repressive Complex 2 Subunit Primary Antibody Unconjugated Public Immunogen Range 1-55/746 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide