Learn More
Bioss KD-Validated LAMP2 Rabbit Reco
SDP
Catalog No. 5000066868
Encompass_Preferred
Lysosomal Associated Membrane Protein 2 Primary Antibody Unconjugated Public Immunogen Range 50-100/362 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66868 Supplier Bioss Supplier No. BSM90523R
Lysosomal Associated Membrane Protein 2 Primary Antibody Unconjugated Public Immunogen Range 50-100/362 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide