Learn More
Bioss KD-Validated LDL receptor rela
SDP
Catalog No. 5000065031
Encompass_Preferred
LDL Receptor Related Protein 8 Primary Antibody Unconjugated Public Immunogen Range 900-963/963 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-65031 Supplier Bioss Supplier No. BSM90117R
LDL Receptor Related Protein 8 Primary Antibody Unconjugated Public Immunogen Range 900-963/963 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide