Learn More
Bioss KD-Validated LPP Rabbit Recomb
SDP
Catalog No. 5000064981
Encompass_Preferred
LIM Domain Containing Preferred Translocation Partner In Lipoma Primary Antibody Unconjugated Public Immunogen Range 1-64/612 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-64981 Supplier Bioss Supplier No. BSM90275R
LIM Domain Containing Preferred Translocation Partner In Lipoma Primary Antibody Unconjugated Public Immunogen Range 1-64/612 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide