Learn More
Bioss KD-Validated LYN Proto-Oncogen
SDP
Catalog No. 5000066822
Encompass_Preferred
LYN Proto-Oncogene Src Family Tyrosine Kinase Primary Antibody Unconjugated Public Immunogen Range 500-555/596 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66822 Supplier Bioss Supplier No. BSM90438R
LYN Proto-Oncogene Src Family Tyrosine Kinase Primary Antibody Unconjugated Public Immunogen Range 500-555/596 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide