Learn More
Bioss KD-Validated Lysine acetyltran
SDP
Catalog No. 5000065183
Encompass_Preferred
Lysine Acetyltransferase 7 Primary Antibody Unconjugated Public Immunogen Range 1-230/611 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-65183 Supplier Bioss Supplier No. BSM90147R
Add to Cart
Add to Cart
Lysine Acetyltransferase 7 Primary Antibody Unconjugated Public Immunogen Range 1-230/611 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide