Learn More
Bioss KD-Validated Lysine demethylas
SDP
Catalog No. 5000065427
Encompass_Preferred
Lysine Demethylase 1B Primary Antibody Unconjugated Public Immunogen Range 75-136/822 Host Species Rabbit Target Species Human Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-65427 Supplier Bioss Supplier No. BSM90258R
Lysine Demethylase 1B Primary Antibody Unconjugated Public Immunogen Range 75-136/822 Host Species Rabbit Target Species Human Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide