Learn More
Bioss KD-Validated Major Vault Prote
SDP
Catalog No. 5000066529
Encompass_Preferred
Major Vault Protein Primary Antibody Unconjugated Public Immunogen Range 1-55/893 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66529 Supplier Bioss Supplier No. BSM90282R
Major Vault Protein Primary Antibody Unconjugated Public Immunogen Range 1-55/893 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide