Learn More
Bioss KD-Validated MAP2K1 Rabbit Rec
SDP
Catalog No. 5000064906
Encompass_Preferred
Mitogen-Activated Protein Kinase Kinase 1 Primary Antibody Unconjugated Public Immunogen Range 1-40/393 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-64906 Supplier Bioss Supplier No. BSM90040R
Mitogen-Activated Protein Kinase Kinase 1 Primary Antibody Unconjugated Public Immunogen Range 1-40/393 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide