Learn More
Bioss KD-Validated MAPK activated pr
SDP
Catalog No. 5000066998
Encompass_Preferred
MAPK Activated Protein Kinase 2 Primary Antibody Unconjugated Public Immunogen Range 650-679/679 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66998 Supplier Bioss Supplier No. BSM90405R
MAPK Activated Protein Kinase 2 Primary Antibody Unconjugated Public Immunogen Range 650-679/679 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide