Learn More
Bioss KD-Validated MAPK10 Rabbit Rec
SDP
Catalog No. 5000067015
Encompass_Preferred
Mitogen-Activated Protein Kinase 10 Primary Antibody Unconjugated Public Immunogen Range 1-36/274 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-67015 Supplier Bioss Supplier No. BSM90432R
Mitogen-Activated Protein Kinase 10 Primary Antibody Unconjugated Public Immunogen Range 1-36/274 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide