Learn More
Bioss KD-Validated Matrix metallopep
SDP
Catalog No. 5000065182
Encompass_Preferred
Matrix Metallopeptidase 14 Primary Antibody Unconjugated Public Immunogen Range 151-182/582 Host Species Rabbit Target Species Human Rat Clonality Monoclonal Applications WB FCM Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-65182 Supplier Bioss Supplier No. BSM90122R
Matrix Metallopeptidase 14 Primary Antibody Unconjugated Public Immunogen Range 151-182/582 Host Species Rabbit Target Species Human Rat Clonality Monoclonal Applications WB FCM Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide