Learn More
ABclonal Technology KD Validated MCT1 Rabbit mAb
SDP
Catalog No. 5000010895
Encompass_Preferred
The protein encoded by this gene is a proton-linked monocarboxylate transporter that catalyzes the movement of many monocarboxylates such as lactate and pyruvate across the plasma membrane Mutations in this gene are associated with erythrocyte lactate transporter defect Alternatively spliced transcript variants have been found for this gene
Catalog No. 50-000-10895 Supplier ABclonal Technology Supplier No. A27270100UL
The protein encoded by this gene is a proton-linked monocarboxylate transporter that catalyzes the movement of many monocarboxylates such as lactate and pyruvate across the plasma membrane Mutations in this gene are associated with erythrocyte lactate transporter defect Alternatively spliced transcript variants have been found for this gene