Learn More
Bioss KD-Validated Mechanistic Targe
SDP
Catalog No. 5000067051
Encompass_Preferred
Mechanistic Target Of Rapamycin Kinase Primary Antibody Unconjugated Public Immunogen Range 1-50/231 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-67051 Supplier Bioss Supplier No. BSM90429R
Mechanistic Target Of Rapamycin Kinase Primary Antibody Unconjugated Public Immunogen Range 1-50/231 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide