Learn More
Bioss KD-Validated Metastasis Associ
SDP
Catalog No. 5000066492
Encompass_Preferred
Metastasis Associated 1 Family Member 2 Primary Antibody Unconjugated Public Immunogen Range 600-668/668 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB FCM Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66492 Supplier Bioss Supplier No. BSM90281R
Metastasis Associated 1 Family Member 2 Primary Antibody Unconjugated Public Immunogen Range 600-668/668 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB FCM Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide