Learn More
Bioss KD-Validated Methionyl aminope
SDP
Catalog No. 5000066878
Encompass_Preferred
Methionyl Aminopeptidase 2 Primary Antibody Unconjugated Public Immunogen Range 420-478/478 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66878 Supplier Bioss Supplier No. BSM90120R
Methionyl Aminopeptidase 2 Primary Antibody Unconjugated Public Immunogen Range 420-478/478 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide