Learn More
Bioss KD-Validated Methylmalonyl-CoA
SDP
Catalog No. 5000064933
Encompass_Preferred
Methylmalonyl-CoA Mutase Primary Antibody Unconjugated Public Immunogen Range 550-580/750 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-64933 Supplier Bioss Supplier No. BSM90195R
Methylmalonyl-CoA Mutase Primary Antibody Unconjugated Public Immunogen Range 550-580/750 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide