Learn More
Bioss KD-Validated Methylthioadenosi
SDP
Catalog No. 5000065188
Encompass_Preferred
Methylthioadenosine Phosphorylase Primary Antibody Unconjugated Public Immunogen Range 201-283/283 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-65188 Supplier Bioss Supplier No. BSM90196R
Methylthioadenosine Phosphorylase Primary Antibody Unconjugated Public Immunogen Range 201-283/283 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide