Learn More
Bioss KD-Validated Mevalonate Kinase
SDP
Catalog No. 5000066272
Encompass_Preferred
Mevalonate Kinase Primary Antibody Unconjugated Public Immunogen Range 320-370/396 Host Species Rabbit Target Species Human Rat Clonality Monoclonal Applications WB IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66272 Supplier Bioss Supplier No. BSM90277R
Mevalonate Kinase Primary Antibody Unconjugated Public Immunogen Range 320-370/396 Host Species Rabbit Target Species Human Rat Clonality Monoclonal Applications WB IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide