Learn More
Bioss KD-Validated Mitochondrial Rib
SDP
Catalog No. 5000065960
Encompass_Preferred
Mitochondrial Ribosomal Protein S15 Primary Antibody Unconjugated Public Immunogen Range 1611-1647/1647 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-65960 Supplier Bioss Supplier No. BSM90427R
Mitochondrial Ribosomal Protein S15 Primary Antibody Unconjugated Public Immunogen Range 1611-1647/1647 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide