Learn More
ABclonal Technology KD Validated MRP4/ABCC4
SDP
Supplier:  ABclonal Technology A2260120UL
Encompass_Preferred
The protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters ABC proteins transport various molecules across extra- and intra-cellular membranes ABC genes are divided into seven distinct subfamilies (ABC1 MDR TAP MRP ALD OABP GCN20 White) This protein is a member of the MRP subfamily which is involved in multi-drug resistance This family member plays a role in cellular detoxification as a pump for its substrate organic anions It may also function in prostaglandin-mediated cAMP signaling in ciliogenesis Alternative splicing of this gene results in multiple transcript variants
Catalog No. 50-000-05215