Learn More
Bioss KD-Validated Musashi RNA Bindi
SDP
Catalog No. 5000066623
Encompass_Preferred
Musashi RNA Binding Protein 2 Primary Antibody Unconjugated Public Immunogen Range 200-280/328 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66623 Supplier Bioss Supplier No. BSM90278R
Musashi RNA Binding Protein 2 Primary Antibody Unconjugated Public Immunogen Range 200-280/328 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide