Learn More
Bioss KD-Validated NDUFS3 Rabbit Rec
SDP
Catalog No. 5000065128
Encompass_Preferred
NADH Primary Antibody Unconjugated Public Immunogen Range 200-264/264 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-65128 Supplier Bioss Supplier No. BSM90023R
NADH Primary Antibody Unconjugated Public Immunogen Range 200-264/264 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide