Learn More
ABclonal Technology KD Validated NNMT Rabbit mAb
SDP
Supplier:  ABclonal Technology A2539120UL
Encompass_Preferred
N-methylation is one method by which drug and other xenobiotic compounds are metabolized by the liver This gene encodes the protein responsible for this enzymatic activity which uses S-adenosyl methionine as the methyl donor
Catalog No. 50-000-11666