Learn More
Bioss KD-Validated Nuclear Mitotic A
SDP
Catalog No. 5000066984
Encompass_Preferred
Nuclear Mitotic Apparatus Protein 1 Primary Antibody Unconjugated Public Immunogen Range 201-265/329 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66984 Supplier Bioss Supplier No. BSM90469R
Nuclear Mitotic Apparatus Protein 1 Primary Antibody Unconjugated Public Immunogen Range 201-265/329 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide