Learn More
Bioss KD-Validated Nucleoporin 155 R
SDP
Catalog No. 5000066833
Encompass_Preferred
Nucleoporin 155 Primary Antibody Unconjugated Public Immunogen Range 901-1050/1391 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66833 Supplier Bioss Supplier No. BSM90010R
Nucleoporin 155 Primary Antibody Unconjugated Public Immunogen Range 901-1050/1391 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide