Learn More
Bioss KD-Validated Olfactomedin 4 Ra
SDP
Catalog No. 5000066988
Encompass_Preferred
Olfactomedin 4 Primary Antibody Unconjugated Public Immunogen Range 1-64/512 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66988 Supplier Bioss Supplier No. BSM90437R
Olfactomedin 4 Primary Antibody Unconjugated Public Immunogen Range 1-64/512 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide