Learn More
Bioss KD-Validated Oxoglutarate rece
SDP
Catalog No. 5000065943
Encompass_Preferred
Oxoglutarate Receptor 1 Primary Antibody Unconjugated Public Immunogen Range 181-215/337 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-65943 Supplier Bioss Supplier No. BSM90198R
Oxoglutarate Receptor 1 Primary Antibody Unconjugated Public Immunogen Range 181-215/337 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide