Learn More
Bioss KD-Validated p38 MAPK Rabbit R
SDP
Catalog No. 5000065942
Encompass_Preferred
Mitogen-Activated Protein Kinase 14 Primary Antibody Unconjugated Public Immunogen Range 150-250/360 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-65942 Supplier Bioss Supplier No. BSM90194R
Mitogen-Activated Protein Kinase 14 Primary Antibody Unconjugated Public Immunogen Range 150-250/360 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide