Learn More
Bioss KD-Validated Paired box 6 Rabb
SDP
Catalog No. 5000066912
Encompass_Preferred
Paired Box 6 Primary Antibody Unconjugated Public Immunogen Range 172-331/422 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66912 Supplier Bioss Supplier No. BSM90125R
Paired Box 6 Primary Antibody Unconjugated Public Immunogen Range 172-331/422 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide