Learn More
Bioss KD-Validated Paraoxonase 2 Rab
SDP
Catalog No. 5000065937
Encompass_Preferred
Paraoxonase 2 Primary Antibody Unconjugated Public Immunogen Range 400-455/455 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB FCM Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-65937 Supplier Bioss Supplier No. BSM90454R
Paraoxonase 2 Primary Antibody Unconjugated Public Immunogen Range 400-455/455 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB FCM Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide