Learn More
Bioss KD-Validated Parkinsonism asso
SDP
Catalog No. 5000064874
Encompass_Preferred
Parkinsonism Associated Deglycase Primary Antibody Unconjugated Public Immunogen Range 1-66/189 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-64874 Supplier Bioss Supplier No. BSM90004R
Parkinsonism Associated Deglycase Primary Antibody Unconjugated Public Immunogen Range 1-66/189 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide