Learn More
Bioss KD-Validated PCBD1 Rabbit Reco
SDP
Catalog No. 5000067056
Encompass_Preferred
Pterin-4 Alpha-Carbinolamine Dehydratase 1 Primary Antibody Unconjugated Public Immunogen Range 481-691/691 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-67056 Supplier Bioss Supplier No. BSM90441R
Pterin-4 Alpha-Carbinolamine Dehydratase 1 Primary Antibody Unconjugated Public Immunogen Range 481-691/691 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide