Learn More
Bioss KD-Validated PCBP2 Rabbit Reco
SDP
Catalog No. 5000065144
Encompass_Preferred
Poly(RC) Binding Protein 2 Primary Antibody Unconjugated Public Immunogen Range 257-300/365 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-65144 Supplier Bioss Supplier No. BSM90020R
Poly(RC) Binding Protein 2 Primary Antibody Unconjugated Public Immunogen Range 257-300/365 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide