Learn More
Bioss KD-Validated Peptidylprolyl Is
SDP
Catalog No. 5000066823
Encompass_Preferred
Peptidylprolyl Isomerase A Primary Antibody Unconjugated Public Immunogen Range 180-224/224 Host Species Rabbit Target Species Human Mouse Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66823 Supplier Bioss Supplier No. BSM90448R
Peptidylprolyl Isomerase A Primary Antibody Unconjugated Public Immunogen Range 180-224/224 Host Species Rabbit Target Species Human Mouse Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide