Learn More
Bioss KD-Validated Peptidylprolyl Is
SDP
Supplier:  Bioss BSM90480R
Encompass_Preferred
Peptidylprolyl Isomerase D Primary Antibody Unconjugated Public Immunogen Range 70-96/271 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-67041