Learn More
Bioss KD-Validated Peptidylprolyl is
SDP
Catalog No. 5000065028
Encompass_Preferred
Peptidylprolyl Isomerase B Primary Antibody Unconjugated Public Immunogen Range 175-216/216 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-65028 Supplier Bioss Supplier No. BSM90002R
Peptidylprolyl Isomerase B Primary Antibody Unconjugated Public Immunogen Range 175-216/216 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide