Learn More
ABclonal Technology KD Validated Peroxiredoxin 6
SDP
Catalog No. 5000010272
Encompass_Preferred
The protein encoded by this gene is a member of the thiol-specific antioxidant protein family This protein is a bifunctional enzyme with two distinct active sites It is involved in redox regulation of the cell it can reduce H(2)O(2) and short chain organic fatty acid and phospholipid hydroperoxides It may play a role in the regulation of phospholipid turnover as well as in protection against oxidative injury
Catalog No. 50-000-10272 Supplier ABclonal Technology Supplier No. A2728920UL
The protein encoded by this gene is a member of the thiol-specific antioxidant protein family This protein is a bifunctional enzyme with two distinct active sites It is involved in redox regulation of the cell it can reduce H(2)O(2) and short chain organic fatty acid and phospholipid hydroperoxides It may play a role in the regulation of phospholipid turnover as well as in protection against oxidative injury