Learn More
Bioss KD-Validated PFKM Rabbit Recom
SDP
Catalog No. 5000065911
Encompass_Preferred
ATP-dependent 6-phosphofructokinase muscle type Primary Antibody Unconjugated Public Immunogen Range 100-150/780 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-65911 Supplier Bioss Supplier No. BSM90127R
ATP-dependent 6-phosphofructokinase muscle type Primary Antibody Unconjugated Public Immunogen Range 100-150/780 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide