Learn More
Bioss KD-Validated Phospho-Paxillin
SDP
Catalog No. 5000065935
Encompass_Preferred
Paxillin Primary Antibody Unconjugated Public Immunogen Range 100-165/165 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-65935 Supplier Bioss Supplier No. BSM90447R
Paxillin Primary Antibody Unconjugated Public Immunogen Range 100-165/165 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide