Learn More
Bioss KD-Validated Phosphoglycerate
SDP
Catalog No. 5000066913
Encompass_Preferred
Phosphoglycerate Kinase 1 Primary Antibody Unconjugated Public Immunogen Range 1-200/417 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66913 Supplier Bioss Supplier No. BSM90128R
Phosphoglycerate Kinase 1 Primary Antibody Unconjugated Public Immunogen Range 1-200/417 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide