Learn More
Bioss KD-Validated PI3 Kinase p110 b
SDP
Catalog No. 5000066624
Encompass_Preferred
Phosphatidylinositol 45-bisphosphate 3-kinase catalytic subunit beta isoform Primary Antibody Unconjugated Public Immunogen Range 5-22/1070 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB FCM Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66624 Supplier Bioss Supplier No. BSM90307R
Phosphatidylinositol 45-bisphosphate 3-kinase catalytic subunit beta isoform Primary Antibody Unconjugated Public Immunogen Range 5-22/1070 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB FCM Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide