Learn More
Bioss KD-Validated Pre-MRNA Processi
SDP
Catalog No. 5000065933
Encompass_Preferred
Pre-MRNA Splicing Tri-SnRNP Complex Factor PRPF4 Primary Antibody Unconjugated Public Immunogen Range 601-685/727 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-65933 Supplier Bioss Supplier No. BSM90451R
Pre-MRNA Splicing Tri-SnRNP Complex Factor PRPF4 Primary Antibody Unconjugated Public Immunogen Range 601-685/727 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide