Learn More
Bioss KD-Validated Proteasome Activa
SDP
Catalog No. 5000066866
Encompass_Preferred
Proteasome Activator Subunit 1 Primary Antibody Unconjugated Public Immunogen Range 250-300/361 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66866 Supplier Bioss Supplier No. BSM90535R
Proteasome Activator Subunit 1 Primary Antibody Unconjugated Public Immunogen Range 250-300/361 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide