Learn More
Bioss KD-Validated PSIP1 / LEDGF Rab
SDP
Catalog No. 5000066820
Encompass_Preferred
PC4 And SRSF1 Interacting Protein 1 Primary Antibody Unconjugated Public Immunogen Range 40-92/637 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66820 Supplier Bioss Supplier No. BSM90444R
PC4 And SRSF1 Interacting Protein 1 Primary Antibody Unconjugated Public Immunogen Range 40-92/637 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide