Learn More
Bioss KD-Validated PTTG1 Rabbit Reco
SDP
Catalog No. 5000067032
Encompass_Preferred
PTTG1 Regulator Of Sister Chromatid Separation Securin Primary Antibody Unconjugated Public Immunogen Range 250-372/372 Host Species Rabbit Target Species Human Mouse Clonality Monoclonal Applications WB FCM Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-67032 Supplier Bioss Supplier No. BSM90476R
PTTG1 Regulator Of Sister Chromatid Separation Securin Primary Antibody Unconjugated Public Immunogen Range 250-372/372 Host Species Rabbit Target Species Human Mouse Clonality Monoclonal Applications WB FCM Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide